L-Adrenalin 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: L-Epinephrine

204400050 204400010 204400250
ACRO204400050EA 114 EUR
ACRO204400050 ACRO204400010 ACRO204400250
L-Adrenalin 99%
L-Adrenaline

Appearance: White to light beige Crystalline powder

Formel: C₉H₁₃NO₃
Molecular Weight: 183,21 g/mol
Schmelzpunkt: 208…211 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002204
CAS-Nummer: 51-43-4
UN: 2811
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 208 to 211 °C
Loss on drying ≤1 % (18 hrs) (0.7 kPa, P2O5)
Sulfated ash ≤0.1 %
Specific optical rotation -50° to -53° (20 °C, 589 nm) (c=4, 1 M HCl) (on dried substance)
Titration with HClO4 ≥98.5 % (On dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR