Vinylmagnesium bromide 0.7 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

209391000 209398000
209391000.EA 54.4 EUR
209391000. 209398000.
Vinylmagnesium bromide 0.7 M in THF, AcroSeal®
Vinylmagnesium bromide

Appearance: Clear yellow to orange to brown Solution

Formel: C₂H₃BrMg
Molecular Weight: 131,26 g/mol
Dichte: 0,98 g/cm³
Flammpunkt: –17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000042
CAS-Nummer: 1826-67-1
UN: 3399
ADR: 4.3,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to orange to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.70 to 0.90 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR