Allyl(chlor)magnesium 1.7 M in THF, AcroSeal®
Lieferant: Thermo Scientific
Gefahr
209671000 209678000
209671000.EA 102 EUR
209671000. 209678000.
Allyl(chlor)magnesium 1.7 M in THF, AcroSeal®
Allyl(chlor)magnesium
Appearance: Clear yellow to green to grey to brown to black Solution
Formel: C₃H₅ClMg
Molecular Weight: 100,83 g/mol
Dichte: 0,96 g/cm³
Flammpunkt: –17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000473
CAS-Nummer: 2622-05-1
UN: 3399
ADR: 4.3,II
Bestellen
Spezifikation Testergebnisse
Appearance Clear yellow to green to grey to brown to black Solution
Titration with HCl 1.5 to 1.9 M