Dicarbonyl(2,4-pentanedionato)rhodium(I)

Lieferant: Thermo Scientific
Gefahr

Synonyme: (Acetylacetonato)dicarbonylrhodium(I), Dicarbonylacetylacetonato rhodium(I)

363955000 363950025
ACRO363955000EA 516 EUR
ACRO363955000 ACRO363950025
Dicarbonyl(2,4-pentanedionato)rhodium(I)
Dicarbonyl(2,4-pentanedionato)rhodium(I)

CAS No.: 14874-82-9

Formel: C₇H₈O₄Rh
Molecular Weight: 259,04 g/mol
Schmelzpunkt: 154…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009884
CAS-Nummer: 14874-82-9
UN: 3467
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Green-red Flakes or needles
Infrared spectrum Conforms
Rhodium (Rh) ≥39.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR