Maltotriose 93%
Lieferant: Thermo Fisher Scientific
Synonyme:
Amylotriose, α-1,4-Glucotriose, α-D-Glc-(1→4)-α-D-Glc-(1→4)-D-Glc, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal
225945000
225940250
ACRO225945000EA
39.2
EUR
ACRO225945000
ACRO225940250
Maltotriose 93%
Maltotriose
Appearance: White Crystalline powder or crystals
Formel:
C₁₈H₃₂O₁₆ Molecular Weight: 504.438 g/mol Schmelzpunkt: 132…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006629 CAS-Nummer: 1109-28-0 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Water | ≤5 % (K.F.) |
HPLC | ≥91 % (ELSD) |
Specific optical rotation | +160° to +165° (25°C, 589 nm) (c=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...