Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (R)-(+)-3-Boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, (R)-(+)-3-(tert-Butoxycarbonyl)-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, Methyl (R)-N-Boc-2,2-dimethyloxazolidine-4-carboxylate, (R)-(+)-2,2-Dimethyloxazolidine-3,4-dicarboxylic acid 3-tert-Butyl 4-Methyl ester, 3-tert-Butyl 4-Methyl (R)-(+)-2,2-Dimethyloxazolidine-3,4-dicarboxylate

426350050 426350010
ACRO426350050EA 196 EUR
ACRO426350050 ACRO426350010
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate 95%
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate

Appearance: Clear colorless to yellow Liquid

Formel: C₁₂H₂₁NO₅
Molecular Weight: 259,302 g/mol
Siedepunkt: 89 °C (,8 mmHg)
Dichte: 1,082 g/cm³
Flammpunkt: 93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00674041
CAS-Nummer: 95715-86-9

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation +53° to +57° (20°C, 589 nm) (C=1, Chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR