Cyclopentylmagnesium chloride 2 M in Diethylether, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

208461000 208468000
208461000.EA 65.2 EUR
208461000. 208468000.
Cyclopentylmagnesium chloride 2 M in Diethylether, AcroSeal®
Cyclopentylmagnesium chloride

Appearance: Clear to hazy yellow to grey to brown Solution

Formel: C₅H₉ClMg
Molecular Weight: 128,882 g/mol
Dichte: 0,86 g/cm³
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001357
CAS-Nummer: 32916-51-1
UN: 3399
ADR: 4.3,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear to hazy yellow to grey to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.9 to 2.2 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR