Bis(4-nitrophenyl)carbonat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bis(4-nitrophenyl) carbonate
171600010
171600050
171600250
ACRO171600010EA
18.3
EUR
ACRO171600010
ACRO171600050
ACRO171600250
Bis(4-nitrophenyl)carbonat 97%
Bis(4-nitrophenyl)carbonat
Formel:
C₁₃H₈N₂O₇ Schmelzpunkt: 139…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007322 CAS-Nummer: 5070-13-3 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Powder |
Infrared spectrum | Conforms |
Melting point | 136 to 144 °C |
GC | ≥96.0 % |
Moisture | 6 % to 10 % |
Bulk density | 475 to 625 kg/m³ |
Acidity | ≤0.1 mg KOH/g (Free Acidity) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...