2-Chlor-4-(trifluormethyl)pyrimidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

397850010 397850050
ACRO397850010EA 60.4 EUR
ACRO397850010 ACRO397850050
2-Chlor-4-(trifluormethyl)pyrimidin 97%
2-Chloro-4-(trifluoromethyl)pyrimidine

Appearance: Clear colorless to light yellow Liquid

Formel: C₅H₂ClF₃N₂
Molecular Weight: 182.5310 g/mol
Siedepunkt: 60 °C (10 mmHg)
Dichte: 1.513 g/cm³
Flammpunkt: 49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00115111
CAS-Nummer: 33034-67-2
UN: 1993
ADR: 3 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR