3,4-Dihydroquinolin-2(1H)-one 98%
Lieferant: Thermo Scientific
|
Achtung
|
398010010
398010100
ACRO398010010EA
40.5
EUR
ACRO398010010
ACRO398010100
3,4-Dihydroquinolin-2(1H)-one 98%
3,4-Dihydroquinolin-2(1H)-one
Appearance: Off-white Crystalline powder
Formel:
C9H9NO Molecular Weight: 147,18 g/mol Schmelzpunkt: 165…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016722 CAS-Nummer: 553-03-7 |
Spezifikation Testergebnisse
Appearance | Off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 164 to 168 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...