4-Aminobenzylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-(Aminomethyl)aniline, alpha-Amino-p-toluidine

391450050 391450250 391451000
ACRO391450050EA 46.8 EUR
ACRO391450050 ACRO391450250 ACRO391451000
4-Aminobenzylamin 98%
4-Aminobenzylamin
Formel: C₇H₁₀N₂
Molecular Weight: 122.17 g/mol
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075513
CAS-Nummer: 4403-71-8
UN: 2735
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Liquid or solid
Infrared spectrum Conforms
Refractive index 1.6080 to 1.6120 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR