(Triphenylphosphin)gold(I)chlorid 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold
391610010
391610050
391612500
ACRO391610010EA
269
EUR
ACRO391610010
ACRO391610050
ACRO391612500
(Triphenylphosphin)gold(I)chlorid 99%
(Triphenylphosphine)gold(I) chloride
Formel:
C₁₈H₁₅AuClP Molecular Weight: 494,71 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00009588 CAS-Nummer: 14243-64-2 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥98.5 % (ex Au) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...