Biphenyl-4-sulfonylchlorid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Biphenyl-4-sulphonyl Chloride

391960050 391960250
ACRO391960050EA 54.2 EUR
ACRO391960050 ACRO391960250
Biphenyl-4-sulfonylchlorid 97%
Biphenyl-4-sulfonylchlorid
Formel: C₁₂H₉ClO₂S
Molecular Weight: 252.72 g/mol
Schmelzpunkt: 114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078658
CAS-Nummer: 1623-93-4
UN: 3261
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 111 to 116 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR