5-Brom-2-nitropyridin 99%
Lieferant: Thermo Scientific
|
Achtung
|
398640010
398640050
ACRO398640010EA
23.2
EUR
ACRO398640010
ACRO398640050
5-Brom-2-nitropyridin 99%
5-Brom-2-nitropyridin
Formel:
C₅H₃BrN₂O₂ Molecular Weight: 203 g/mol Schmelzpunkt: 148…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160411 CAS-Nummer: 39856-50-3 |
Spezifikation Testergebnisse
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 148 to 152 °C |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...