[(2-Bromethoxy)methyl]benzol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: ((2-Bromoethoxy)methyl)benzene, Benzyloxyethyl Bromide, (2-Bromoethoxymethyl)benzene

388870050 388870250 388871000
ACRO388870050EA 129 EUR
ACRO388870050 ACRO388870250 ACRO388871000
[(2-Bromethoxy)methyl]benzol 97%
[(2-Bromethoxy)methyl]benzol

Appearance: Clear colorless to light yellow Liquid

Formel: C₉H₁₁BrO
Molecular Weight: 215.09 g/mol
Siedepunkt: 254 °C (760 mmHg)
Dichte: 1.36 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD01321307
CAS-Nummer: 1462-37-9
UN: 3334
ADR: 9 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5420 (20°C, 589 nm)
GC ≥96.0 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR