2-Methyl-2-propanyl-(2-bromethyl)carbamat 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(BOC-amino)ethyl bromide

398940010 398940050
ACRO398940010EA 54.7 EUR
ACRO398940010 ACRO398940050
2-Methyl-2-propanyl-(2-bromethyl)carbamat 96%
tert-Butyl (2-bromoethyl)carbamate

Appearance: Off-white to beige Low melting solid

Formel: C₇H₁₄BrNO₂
Molecular Weight: 224.10 g/mol
Schmelzpunkt: 30…32 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD02683428
CAS-Nummer: 39684-80-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to beige Low melting solid
Infrared spectrum Conforms
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR