3-Methoxybenzyl bromide 98+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
393110050
393110250
ACRO393110050EA
83.2
EUR
ACRO393110050
ACRO393110250
3-Methoxybenzyl bromide 98+%
3-Methoxybenzyl bromide
Appearance: Clear colorless to light yellow Liquid
Formel:
C₈H₉BrO Siedepunkt: 152 °C (760 mmHg) Dichte: 1,436 g/cm³ Flammpunkt: 113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216590 CAS-Nummer: 874-98-6 UN: 3265 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5740 to 1.5760 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...