2'-Iodacetophenon 99+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(2-Iodphenyl)ethanon, o-Iodacetophenon, 1-(2-Iodphenyl)ethan-1-on
393130010
393130050
ACRO393130010EA
32.2
EUR
ACRO393130010
ACRO393130050
2'-Iodacetophenon 99+%
2'-Iodacetophenon
Formel:
C₈H₇IO Molecular Weight: 246,05 g/mol Siedepunkt: 139…140 °C (12 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00094998 CAS-Nummer: 2142-70-3 UN: 2810 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6170 to 1.6200 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...