Chromium(III) chloride tetrahydrofuran complex (1:3) 98%
Lieferant: Thermo Scientific
Achtung
389850010 389850100
ACRO389850010EA 38.4 EUR
ACRO389850010 ACRO389850100
Chromium(III) chloride tetrahydrofuran complex (1:3) 98%
Chromium(III) chloride tetrahydrofuran complex (1:3)
Appearance: Purple Crystalline powder
Formel: C₁₂H₂₄Cl₃CrO₃
Molecular Weight: 374,667 g/mol
Schmelzpunkt: 143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013222
CAS-Nummer: 10170-68-0
UN: 1325
ADR: 4.1,II
Bestellen
Spezifikation Testergebnisse
Appearance Purple Crystalline powder
Infrared spectrum Conforms
Titration with Na2S2O3 ≥97.5 %