4-Methoxypiperidin 98+%
Lieferant: Thermo Scientific
|
Gefahr
|
393390010
393390050
ACRO393390010EA
128
EUR
ACRO393390010
ACRO393390050
4-Methoxypiperidin 98+%
4-Methoxypiperidin
Formel:
C₆H₁₄ClNO Molecular Weight: 115,1700 g/mol Siedepunkt: 44 °C (9mbar) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115010 CAS-Nummer: 4045-24-3 UN: 3267 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4590 to 1.4610 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...