2-Chlor-5-fluor-4-pyrimidinamin 98%

Lieferant: Thermo Scientific
Achtung

399980010 399980050
ACRO399980010EA 216 EUR
ACRO399980010 ACRO399980050
2-Chlor-5-fluor-4-pyrimidinamin 98%
2-Chlor-5-fluor-4-pyrimidinamin

Appearance: White to light yellow Crystalline powder

Formel: C₄H₃ClFN₃
Molecular Weight: 147,54 g/mol
Schmelzpunkt: 193…195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00057344
CAS-Nummer: 155-10-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 192 to 196 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR