Lithium isopropoxide 3 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
396501000
396501000.EA
101
EUR
396501000.
Lithium isopropoxide 3 M in THF, AcroSeal®
Lithium isopropoxide
Appearance: Clear red Solution
Formel:
C₃H₇LiO Molecular Weight: 66.028 g/mol Dichte: 0.87 g/cm³ Flammpunkt: –19 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048286 CAS-Nummer: 2388-10-5 UN: 2924 ADR: 3 II |
Spezifikation Testergebnisse
Appearance | Clear yellow to red Solution |
Strength | 2.9 to 3.3 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...