5-Formyl-2-thiophenboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid

397150010 397150050
ACRO397150010EA 72 EUR
ACRO397150010 ACRO397150050
5-Formyl-2-thiophenboronsäure 97%
5-Formyl-2-thiophenboronsäure
Formel: C₅H₅BO₃S
Molecular Weight: 155,97 g/mol
Schmelzpunkt: 132…135 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02093666
CAS-Nummer: 4347-33-5

Bestellen

Spezifikation Testergebnisse

Appearance White to orange Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR