2-Ethylbenzoesäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
378750010
378750050
ACRO378750010EA
110
EUR
ACRO378750010
ACRO378750050
2-Ethylbenzoesäure 97%
2-Ethylbenzoic acid
Formel:
C₉H₁₀O₂ Molecular Weight: 150.1700 g/mol Schmelzpunkt: 62…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045838 CAS-Nummer: 612-19-1 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 62 to 66 °C |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...