(tert-Butyldimethylsiloxy)acetaldehyde 90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (tert-Butyldimethylsilyloxy)acetaldehyde

397510010 397510050
ACRO397510010EA 79.7 EUR
ACRO397510010 ACRO397510050
(tert-Butyldimethylsiloxy)acetaldehyde 90%
(tert-Butyldimethylsiloxy)acetaldehyde

Appearance: Clear colorless to straw yellow Liquid

Formel: C₈H₁₈O₂Si
Molecular Weight: 174.315 g/mol
Siedepunkt: 156…157 °C (760 mmHg)
Dichte: 0.915 g/cm³
Flammpunkt: 60 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01321229
CAS-Nummer: 102191-92-4
UN: 1989
ADR: 3 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to straw yellow Liquid
Infrared spectrum Conforms
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR