1-Boc-4-(hydroxymethyl)piperidin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol
397720010
397720050
ACRO397720010EA
15.7
EUR
ACRO397720010
ACRO397720050
1-Boc-4-(hydroxymethyl)piperidin 97%
1-Boc-4-(hydroxymethyl)piperidin
Formel:
C₁₁H₂₁NO₃ Molecular Weight: 215,29 g/mol Schmelzpunkt: 78…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02094488 CAS-Nummer: 123855-51-6 |
Spezifikation Testergebnisse
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...