ω-Brom-2-chloracetophenon 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Brom-2'-chloracetophenon, 2-Chlorphenacylbromid, Brommethyl-o-chlorphenylketon, α-Brom-2-chloracetophenon
427890010
427890050
ACRO427890010EA
64.6
EUR
ACRO427890010
ACRO427890050
ω-Brom-2-chloracetophenon 95%
ω-Brom-2-chloracetophenon
Formel:
C₈H₆BrClO Molecular Weight: 233,49 g/mol Siedepunkt: 162…166 °C (20 mmHg) Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00832993 CAS-Nummer: 5000-66-8 UN: 3265 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
NMR (1H-NMR) | Consistent with structure |
Chloride (Cl) | ≤0.001 % |
Oxalate (C2O4) | (limit about 0.05%) Passes test |
Phosphate (PO4) | ≤0.001 % |
Sulfate (SO4) | ≤0.002 % |
Iron (Fe) | ≤3 ppm |
Lead (Pb) | ≤2 ppm |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...