cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid 95%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: cis-2-Carboxy-3-azabicyclo[3.1.0]hexane
425900010 425902500
ACRO425900010EA 379 EUR
ACRO425900010 ACRO425902500
cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid 95%
cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid
Appearance: White to light yellow Crystalline powder
Formel: C₆H₉NO₂
Schmelzpunkt: 252…256 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03093875
CAS-Nummer: 22255-16-9
Bestellen
Spezifikation Testergebnisse
Appearance White to yellow Powder or crystalline powder
Infrared spectrum Conforms
NMR (1H-NMR) Consistent with structure
Titration with NaOH ≥94 %