(6-Brom-2-pyridinyl)methanol 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: (6-Bromopyridin-2-yl)methanol, 2-Bromo-6-pyridinemethanol, 2-Bromo-6-(hydroxymethyl)pyridine

426310010 426310050
ACRO426310010EA 61.5 EUR
ACRO426310010 ACRO426310050
(6-Brom-2-pyridinyl)methanol 96%
(6-Brom-2-pyridinyl)methanol
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
Schmelzpunkt: 34…39 °C
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039884
CAS-Nummer: 33674-96-3
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 34 to 41 °C
GC ≥95 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR