1-Methyl-1H-indole-3-carboxylic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Methylindole-3-carboxylic acid

426480010 426480050
ACRO426480010EA 24.3 EUR
ACRO426480010 ACRO426480050
1-Methyl-1H-indole-3-carboxylic acid 97%
1-Methyl-1H-indole-3-carboxylic acid

Appearance: White to beige Crystalline powder

Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Schmelzpunkt: 197…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01321244
CAS-Nummer: 32387-21-6

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder or crystalline powder
Infrared spectrum Conforms
Melting point 197 to 202 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR