(3R)-(+)-3-(Dimethylamino)pyrrolidine 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (R)-(+)-3-(Dimethylamino)pyrrolidine, (R)-N,N-Dimethylpyrrolidin-3-amine

426990010 426990050
ACRO426990010EA 108 EUR
ACRO426990010 ACRO426990050
(3R)-(+)-3-(Dimethylamino)pyrrolidine 98%
(3R)-(+)-3-(Dimethylamino)pyrrolidine

Appearance: Clear colorless to yellow Liquid

Formel: C₆H₁₄N₂
Molecular Weight: 114.19 g/mol
Siedepunkt: 166 °C (760 mmHg)
Flammpunkt: 51.7 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191347
CAS-Nummer: 132958-72-6
UN: 2733
ADR: 3 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation +12° to +16° (20°C, 589 nm) (c=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR