trans-N1,N2-Dimethylcyclohexane-1,2-diamine

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: trans-N,N'-Dimethyl-1,2-cyclohexanediamine

427310050 427310250
ACRO427310050EA 208 EUR
ACRO427310050 ACRO427310250
trans-N1,N2-Dimethylcyclohexane-1,2-diamine
trans-N1,N2-Dimethylcyclohexane-1,2-diamine

Appearance: Clear colorless to light yellow Liquid

Formel: C₈H₁₈N₂
Molecular Weight: 142.24 g/mol
Siedepunkt: 78…80 °C (18 mmHg)
Dichte: 0.902 g/cm³
Flammpunkt: 74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03001702
CAS-Nummer: 67579-81-1
UN: 2735
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4730 (20°C, 589 nm)
GC ≥96.0 %
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.0001 meq/g
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.2 ppm
Potassium (K) ≤0.5 ppm
Sodium (Na) ≤0.5 ppm
Zinc (Zn) ≤0.1 ppm
Impurity ≤1 % n-Propyl benzene
Benzene ≤0.05 %
KMnO4-reducing substance ≤5 ppm (O)
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR