3-Butenylmagnesium bromide 0.5 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

427461000 427468000
427461000.EA 401 EUR
427461000. 427468000.
3-Butenylmagnesium bromide 0.5 M in THF, AcroSeal®
3-Butenylmagnesium bromide

Appearance: Grey to brown Solution

Formel: C₄H₇BrMg
Molecular Weight: 159,309 g/mol
Siedepunkt: 65 °C (760 mmHg)
Dichte: 0,937 g/cm³
Flammpunkt: –30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00209554
CAS-Nummer: 7103-09-5
UN: 3399
ADR: 4.3,I

Bestellen

Spezifikation Testergebnisse

Appearance Grey to brown Solution
Titration Grignard 0.40 to 0.60 M
Identification Confirmed

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR