4-Brom-1-indanon 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-bromo-2,3-dihydro-1H-inden-1-one
427700010
427700050
ACRO427700010EA
39.9
EUR
ACRO427700010
ACRO427700050
4-Brom-1-indanon 97%
4-Bromo-1-indanone
Appearance: Orange-brown Crystalline powder
Formel:
C₉H₇BrO Molecular Weight: 211,06 g/mol Schmelzpunkt: 95…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01719772 CAS-Nummer: 15115-60-3 |
Spezifikation Testergebnisse
Appearance | Orange-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 94 to 98 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...