Acroleindiethylacetal 94%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Acroleinacetal, 3,3-Diethoxypropen

102770250 102771000
ACRO102770250EA 57.6 EUR
ACRO102770250 ACRO102771000
Acroleindiethylacetal 94%
Acroleindiethylacetal
Formel: C₇H₁₄O₂
Molecular Weight: 130,19 g/mol
Siedepunkt: 120…125 °C (760 mmHg)
Schmelzpunkt: –77 °C
Dichte: 0,85 g/cm³
Flammpunkt: 15 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009239
CAS-Nummer: 3054-95-3
UN: 2374
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.4030 (20°C, 589 nm)
GC ≥92 %
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Sulfur compounds (as SO4) ≤0.002 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR