Cholesteryl-benzoat 98%

Lieferant: Thermo Scientific

Synonyme: Cholesteryl benzoate, 3b-(Benzoyloxy)cholest-5-ene

110200250 110201000
ACRO110200250EA 62.6 EUR
ACRO110200250 ACRO110201000
Cholesteryl-benzoat 98%
Cholesterol benzoate
Formel: C₃₄H₅₀O₂
Molecular Weight: 490,77 g/mol
Schmelzpunkt: 149…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003635
CAS-Nummer: 604-32-0

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration RCOCl ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR