2-Nitrobenzolsulfonylchlorid 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Nitrobenzolsulfonsäurechlorid, 2-Nitrobenzol-1-sulfonylchlorid, 2-Nitrobenzolsulfochlorid

128430250 128431000
ACRO128430250EA 38.7 EUR
ACRO128430250 ACRO128431000
2-Nitrobenzolsulfonylchlorid 97%
2-Nitrobenzolsulfonylchlorid
Formel: C₆H₄ClNO₄S
Molecular Weight: 221,62 g/mol
Schmelzpunkt: 63…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007430
CAS-Nummer: 1694-92-4
UN: 3261
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance White to cream to yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 64 to 70 °C
GC ≥96.0 %
Enantiomeric excess ≥99.0 %
Specific optical rotation +53° to +59° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR