4-Nitroanthranilsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-nitrobenzoic acid
156980050
156980250
ACRO156980050EA
230
EUR
ACRO156980050
ACRO156980250
4-Nitroanthranilsäure 97%
4-Nitroanthranilsäure
Formel:
C₇H₆N₂O₄ Molecular Weight: 182,14 g/mol Schmelzpunkt: 268…269 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007262 CAS-Nummer: 619-17-0 |
Spezifikation Testergebnisse
Appearance | Orange Crystalline powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...