1-Methyl-1H-indol-2-carbonsäure
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Indole-2-carboxylic acid, 1-methyl-, 1-Methylindole-2-carboxylic acid
158640010
158640050
ACRO158640010EA
58.4
EUR
ACRO158640010
ACRO158640050
1-Methyl-1H-indol-2-carbonsäure
1-Methyl-1H-indole-2-carboxylic acid
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Schmelzpunkt: 212…213 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005801 CAS-Nummer: 16136-58-6 |
Spezifikation Testergebnisse
Appearance | Beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Sulfate (SO4) | ≤0.005 % |
Arsenic (As) | ≤0.001 % |
Calcium (Ca) | ≤0.005 % |
Copper (Cu) | ≤0.002 % |
Iron (Fe) | ≤0.001 % |
Lead (Pb) | ≤0.002 % |
Potassium (K) | ≤0.01 % |
Silver (Ag) | ≤0.001 % |
Sodium (Na) | ≤0.02 % |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...