Quinoxaline-2-carbonyl chloride 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Quinoxaloyl chloride
163760010
163762500
ACRO163760010EA
161
EUR
ACRO163760010
ACRO163762500
Quinoxaline-2-carbonyl chloride 95%
Quinoxaline-2-carbonyl chloride
Formel:
C₉H₅ClN₂O Molecular Weight: 192,61 g/mol Schmelzpunkt: 113…115 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006725 CAS-Nummer: 54745-92-5 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Insoluble matter | ≤0.05 % (in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...