1-Isochinolinol 98%

Lieferant: Thermo Scientific

Synonyme: Isocarbostyril, 1-Hydroxyisoquinoline

166460010 166460050
ACRO166460010EA 59.7 EUR
ACRO166460010 ACRO166460050
1-Isochinolinol 98%
1-Isochinolinol
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
Schmelzpunkt: 211…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006899
CAS-Nummer: 491-30-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 210 to 215 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR