4-(4-Methoxyphenyl)butansäure 98%

Lieferant: Thermo Scientific
Achtung

172440050
ACRO172440050EA 56.9 EUR
ACRO172440050
4-(4-Methoxyphenyl)butansäure 98%
4-(4-Methoxyphenyl)butansäure
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
Schmelzpunkt: 56…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004404
CAS-Nummer: 4521-28-2

Bestellen

Spezifikation Testergebnisse

Appearance Cream to light yellow Powder
Infrared spectrum Conforms
Melting point 56 to 61 °C
Titration with TBAH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR