3-Chinolincarbaldehyd 97+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Chinolincarboxaldehyd

175690010 175690050
ACRO175690010EA 98.8 EUR
ACRO175690010 ACRO175690050
3-Chinolincarbaldehyd 97+%
3-Chinolincarbaldehyd
Formel: C₁₀H₇NO
Molecular Weight: 157,17 g/mol
Schmelzpunkt: 66…69 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006768
CAS-Nummer: 13669-42-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 66 to 69 °C
GC ≥97.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR