2-(3-Methoxyphenyl)ethanol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(3-Methoxyphenyl)ethylalkohol, 3-Methoxyphenethylalkohol

182250010 182250050 182250250
ACRO182250010EA 48.7 EUR
ACRO182250010 ACRO182250050 ACRO182250250
2-(3-Methoxyphenyl)ethanol 97%
2-(3-Methoxyphenyl)ethanol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Siedepunkt: 141…143 °C (12 mmHg)
Dichte: 1,075 g/cm³
Flammpunkt: >113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002893
CAS-Nummer: 5020-41-7

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure
Titration with HClO4 97.5 to 102.5 % (dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR