L-Leucic acid 98%

Lieferant: Thermo Fisher Scientific
Gefahr

204810010 204810050
ACRO204810010EA 46 EUR
ACRO204810010 ACRO204810050
L-Leucic acid 98%
L-Leucic acid
Formel: C₆H₁₂O₃
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064214
CAS-Nummer: 13748-90-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 81 °C
NMR consistent with structure
Specific optical rotation -24° to -28° (20 °C, 589 nm) (C=1, 1N NaOH)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR