(4-Bromophenyl)(cyclopropyl)ketone 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Bromophenyl)(cyclopropyl)methanone, (4-Bromophenyl)cyclopropylmethanone, (4-Bromobenzoyl)cyclopropane, 4-Bromophenyl Cyclopropyl ketone
235420500
235422500
ACRO235420500EA
178
EUR
ACRO235420500
ACRO235422500
(4-Bromophenyl)(cyclopropyl)ketone 95%
(4-Bromophenyl)(cyclopropyl)ketone
Formel:
C₁₀H₉BrO Molecular Weight: 225,09 g/mol Dichte: 1,45 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019227 CAS-Nummer: 6952-89-2 |
Spezifikation Testergebnisse
Appearance | Yellow to beige to orange Crystalline powder or low melting crystalline mass |
Infrared spectrum | Conforms |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...