5-Brom-2-thenoesäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Bromthiophen-2-carbonsäure, 5-Brom-2-thiophencarbonsäure
243640010
243640050
ACRO243640010EA
24.1
EUR
ACRO243640010
ACRO243640050
5-Brom-2-thenoesäure 97%
5-Brom-2-thenoesäure
Formel:
C₅H₃BrO₂S Molecular Weight: 207.05 g/mol Schmelzpunkt: 140…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079725 CAS-Nummer: 7311-63-9 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 141 to 145 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...