Sodium-tert-pentyloxide 30% in 2-MeTHF (2-Methyltetrahydrofuran), AcroSeal®
Lieferant: Thermo Scientific
Gefahr
Synonyme: Sodium tert-pentoxide, Sodium tert-pentoxide, 95%, tert-Amyl alcohol sodium salt
252531000 252538000
252531000.EA 75 EUR
252531000. 252538000.
Sodium-tert-pentyloxide 30% in 2-MeTHF (2-Methyltetrahydrofuran), AcroSeal®
Sodium-tert-pentyloxide
Formel: C₅H₁₁NaO
Molecular Weight: 110,132 g/mol
Dichte: 0,91 g/cm³
Flammpunkt: –11 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064245
CAS-Nummer: 14593-46-5
UN: 2924
ADR: 3,II
Bestellen
Spezifikation Testergebnisse
Appearance Yellow to orange to brown Solution (May develop some turbidity or precipitate)
Titration with HClO4 ≥30 %