L-(-)-α-Amino-γ-butyrolactone hydrobromide 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
L-(-)-α-Amino-γ-butyrolactone hydrobromide, (S)-2-amino-4-butyrolactone hydrobromide, (S)-(-)-α-Amino-γ-butyrolactone hydrobromide, (S)-3-Aminodihydrofuran-2(3H)-one hydrobromide
259940010
259940050
ACRO259940010EA
37.2
EUR
ACRO259940010
ACRO259940050
L-(-)-α-Amino-γ-butyrolactone hydrobromide 99%
L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide
Formel:
C₄H₇NO₂·HBr Molecular Weight: 182,017 g/mol Schmelzpunkt: 236…238,9 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674493 CAS-Nummer: 15295-77-9 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
Specific optical rotation | -20.5° to -22.5° (20°C, 589 nm) (c=4, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...