N-Methylphthalimid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-Methylphthalsäureimid, N-Methyl-1,2-benzoldicarbonsäureimid
305100050
305100250
305101000
ACRO305100050EA
20.8
EUR
ACRO305100050
ACRO305100250
ACRO305101000
N-Methylphthalimid 98%
N-Methylphthalimid
Formel:
C₉H₇NO₂ Molecular Weight: 161,16 g/mol Siedepunkt: 286 °C (1013 hPa) Schmelzpunkt: 129…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023063 CAS-Nummer: 550-44-7 |
Spezifikation Testergebnisse
Appearance | White to almost white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 132 to 136 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...